ContactUsLinkComponent
Human ST8SIA2 ELISA Kit
Catalog # ABCAAB314356-96TST
Supplier:  Abcam
CAS Number:  
undefined
Human ST8SIA2 ELISA Kit
Catalog # ABCAAB314356-96TST
Supplier:  Abcam
CAS Number:  
About this item
Human ST8SIA2 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human ST8SIA2 in Human in cell culture supernatant, plasma, serum samples.