Adalimumab ELISA Kit
Catalog # ABCAAB237641-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Adalimumab ELISA kit
Specifications
About this item
Adalimumab ELISA kit ab237641 is used to quantify Adalimumab with a sensitivity of 10 ng/ml. Biosimilar ELISA kit.