Human TAFA1 ELISA Kit
Catalog # ABCAAB314359-96TST
Supplier: Abcam
CAS Number:
Specifications
- Description:Human TAFA1 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human TAFA1 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human TAFA1 in Human in Cell culture supernatant, Plasma, Serum samples.