Human IgA ELISA Kit
Human IgA ELISA kit is a single-wash 90-min SimpleStep ELISA used to quantification of human Immunoglobulin A (IgA) with a sensitivity of 0,25 ng/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric Sandwich ELISA - 450 nm readout; works on any plate reader. Different formats for different needs: 384-well for higher throughput. Broad sample compatibility - works with saliva urine, plasma, cell culture supernatants, milk, and serum.
Human IgA ELISA kit is a single-wash 90-min SimpleStep ELISA used to quantification of human Immunoglobulin A (IgA) with a sensitivity of 0,25 ng/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric Sandwich ELISA - 450 nm readout; works on any plate reader. Different formats for different needs: 384-well for higher throughput. Broad sample compatibility - works with saliva urine, plasma, cell culture supernatants, milk, and serum.
- Catalog No:
- ABCAAB196263-96
- ABCAAB196263-1X384
- Target protein:
- IgA
- IgA
- Primary antibody reactivity:
- Human
- Human
- Assay Type:
- Sandwich
- Sandwich
- Format:
- Pre-coated
- Pre-coated
- Assay duration:
- One step
- One step
- Sensitivity:
- 0,25 ng/ml
- 0,25 ng/ml
- Detection range:
- 0,78 - 50 ng/ml
- 0,78 - 50 ng/ml
- Detection method:
- Colorimetric
- Colorimetric
- Sample type:
- Saliva, Urine, Plasma, Cell culture supernatant, Milk, Serum
- Saliva, Urine, Plasma, Cell culture supernatant, Milk, Serum
- Time to Results:
- 1 h 30 min
- 1 h 30 min
- Regulatory status:
- RUO
- RUO
- Storage temperature:
- 4 ℃
- 4 ℃
Product Family Options
- DescriptionTests per kit
Specifications
Cat. No.ABCAAB196263-96Target proteinIgAPrimary antibody reactivityHumanAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity0,25 ng/mlDetection range0,78 - 50 ng/mlDetection methodColorimetricSample typeSaliva, Urine, Plasma, Cell culture supernatant, Milk, SerumTime to Results1 h 30 minRegulatory statusRUOStorage temperature4 ℃- Human IgA ELISA kit384 tests1 * 384 Tests1 930,00 €
Specifications
Cat. No.ABCAAB196263-1X384Target proteinIgAPrimary antibody reactivityHumanAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity0,25 ng/mlDetection range0,78 - 50 ng/mlDetection methodColorimetricSample typeSaliva, Urine, Plasma, Cell culture supernatant, Milk, SerumTime to Results1 h 30 minRegulatory statusRUOStorage temperature4 ℃



