Human IL-29 ELISA Kit
ABCAAB100568-96
: Abcam
:
- Description:Human IL-29 ELISA kit
- Tests per kit:96 tests
Human IL-29 ELISA kit ab100568 is used to quantify Human IL-29 with a sensitivity of 20 pg/ml. Sandwich ELISA kit.
Human IL-29 ELISA kit ab100568 is used to quantify Human IL-29 with a sensitivity of 20 pg/ml. Sandwich ELISA kit.