Human LAG3 ELISA Kit
ABCAAB193707-96
: Abcam
:
- Description:Human LAG3 ELISA Kit
- Tests per kit:96 tests
Human LAG3 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human LAG3 in Human in Plasma, Cell culture supernatant, Serum samples.