Human PMP2 ELISA Kit
ABCAAB313599-96TST
: Abcam
:
- Description:Human PMP2 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Human PMP2 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human PMP2 in Human in cell culture supernatant, plasma, serum samples.