ContactUsLinkComponent
Phosphotyrosine Fyn ELISA Kit
Phosphotyrosine Fyn ELISA Kit
Catalog # ABCAAB279807-1X96
Supplier:  Abcam
CAS Number:  
undefined
Phosphotyrosine Fyn ELISA Kit
Catalog # ABCAAB279807-1X96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Phosphotyrosine Fyn ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
Phosphotyrosine Fyn ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine Fyn in Human in Cell Lysate samples.