Mouse IL-33 ELISA Kit
ABCAAB213475-96
: Abcam
:
- Description:Mouse IL-33 ELISA kit
Mouse IL-33 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse IL-33 with a sensitivity of 5.3 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.