Rat RANK ELISA Kit
Catalog # ABCAAB213918-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat RANK ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Rat RANK ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat RANK in Rat in cell culture supernatant, serum samples.