Human GLP-1 (7-36) ELISA Kit, Fluorescent
ABCAAB229413-1X96
: Abcam
:
- Description:Human GLP-1 (7-36) ELISA kit, Fluorescent
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Human GLP-1 (7-36) ELISA kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Human GLP-1 (7-36) with a sensitivity of 12 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Fluorescent Sandwich ELISA - 530/570/590 nm readout: works on any standard plate reader.