Arg8-Vasopressin ELISA Kit
ABCAAB205928-96
: Abcam
:
- Description:Arg8-Vasopressin ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Arg8-Vasopressin ELISA kit is a Competitive ELISA kit for the measurement of Arg8-Vasopressin in plasma, cell culture supernatant, serum samples.