Human Elastin ELISA Kit
About this item
Human Elastin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Elastin with a sensitivity of 44 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.
2 Options Available Below
Product Details & Documents
Human Elastin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Elastin with a sensitivity of 44 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ABCAAB239433-96
- ABCAAB239433-1X384
- Target protein:
- Elastin
- Elastin
- Primary antibody reactivity:
- Human
- Human
- Assay Type:
- Sandwich
- Sandwich
- Format:
- Pre-coated
- Pre-coated
- Assay duration:
- One step
- One step
- Sensitivity:
- 44 pg/ml
- 44 pg/ml
- Detection range:
- 0,19 - 12 ng/ml
- 0,19 - 12 ng/ml
- Detection method:
- Colorimetric
- Colorimetric
- Sample type:
- Citrate plasma, Serum, EDTA Plasma
- Citrate plasma, Serum, EDTA Plasma
- Time to Results:
- 1 h 30 min
- 1 h 30 min
- Regulatory status:
- RUO
- RUO
- Storage temperature:
- +4 °C
- +4 °C
Specifications
Product Family Options
Product Information
- Tests per kitAvailabilityPrice
- ABCAAB239433-96Human Elastin ELISA kitSpecifications:
More
Specifications
Cat. No.ABCAAB239433-96Target proteinElastinPrimary antibody reactivityHumanAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity44 pg/mlDetection range0,19 - 12 ng/mlDetection methodColorimetricSample typeCitrate plasma, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOStorage temperature+4 °C - ABCAAB239433-1X384Human Elastin ELISA kitSpecifications:New Product
More
Specifications
Cat. No.ABCAAB239433-1X384Target proteinElastinPrimary antibody reactivityHumanAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity44 pg/mlDetection range0,19 - 12 ng/mlDetection methodColorimetricSample typeCitrate plasma, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOStorage temperature+4 °CNew Product



