Lysozyme ELISA Kit
Lysozyme ELISA Kit, 96 tests
Catalog # ABCAAB285338-96
Supplier: Abcam
Jump to:
Product Details & Documents
Lysozyme ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Lysozyme in Human in serum, Other biological fluids samples.
Specifications
Specifications
- Description:Lysozyme ELISA Kit
- Tests per kit:96 tests
- Cat. No.:ABCAAB285338-96
- Assay duration:Multiple steps
- Format:Pre-coated
- Primary antibody reactivity:Human
- Sample type:Serum, Other biological fluids
- Target protein:Lysozyme
- Storage temperature:4 °C
- Regulatory status:RUO
- Assay Type:Sandwich



