Human CRHBP ELISA Kit
ABCAAB313676-96TST
: Abcam
:
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:CRHBP
- Description:Human CRHBP ELISA kit
- Sample type:Cell culture supernatant, plasma, serum
- Detection method:Colorimetric
- Detection range:0,07 - 20 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,07 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Human CRHBP ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human CRHBP in Human in cell culture supernatant, plasma, Serum samples.