Human LILRA3 ELISA Kit
ABCAAB313983-96TST
: Abcam
:
- Description:Human LILRA3 ELISA Kit
- Tests per kit:96 tests
Human LILRA3 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human LILRA3 in Human in Serum, Plasma, Cell culture supernatant samples.