Human GAP43 ELISA Kit
Catalog # ABCAAB282867-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human GAP43 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human GAP43 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human GAP43 with a sensitivity of 5,9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.