Estrogen ELISA Kit
Catalog # ABCAAB285313-96
Supplier: Abcam
CAS Number:
Specifications
- Pk:96 Tests
- Assay Type:Competitive
- Description:Estrogen ELISA kit
- Tests per kit:100 assays
Specifications
About this item
Estrogen ELISA kit is a Quantitative ELISA kit for the measurement of Estrogen in plasma, Tissue Homogenate, serum, Other biological fluids samples.
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.