Human Granzyme B ELISPOT Kits
Supplier: Abcam
Certificates
About this item
Human Granzyme B ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Human Granzyme B production in a single cell suspension.
Specifications
- Species reactivity:Human
- Form:Pre-coated
- Antigen name:Granzyme B
- Storage temperature:4 °C
- Processing time:Multiple steps
- Regulatory status:RUO