Mouse IL-18 ELISA Kit
Catalog # ABCAAB216165-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse IL-18 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse IL-18 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse IL-18 with a sensitivity of 10,5 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput. - Cited in over 70 citations.