Human IP-10 ELISA Kit
Supplier: Abcam
Certificates
About this item
Human IP-10 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IP-10 with a sensitivity of 1,039 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.