Mouse CXCL16 ELISA Kit
Catalog # ABCAAB100677-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse CXCL16 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse CXCL16 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse CXCL16 in Mouse in plasma, cell culture supernatant, serum samples.