Protein A ELISA Kit
Catalog # ABCAAB133036-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Protein A ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Protein A ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Protein A.