ContactUsLinkComponent
Hamster CHO MMP-19 ELISA Kit
Hamster CHO MMP-19 ELISA Kit
Catalog # ABCAAB313942-96TST
Supplier:  Abcam
New Product
CAS Number:  
undefined
Hamster CHO MMP-19 ELISA Kit
Catalog # ABCAAB313942-96TST
Supplier:  Abcam
New Product
CAS Number:  
Specifications
  • Description:
    Hamster CHO MMP-19 ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Hamster CHO MMP-19 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Hamster CHO MMP-19 in Chinese hamster in Tissue Culture Media samples.