Mouse Interferon alpha 1 ELISA Kit
About this item
Mouse Interferon alpha 1 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Interferon alpha 1 with a sensitivity of 7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 10 citations.
3 Options Available Below
Product Details & Documents
Mouse Interferon alpha 1 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Interferon alpha 1 with a sensitivity of 7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 10 citations.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ABCAAB252352-1X96
- ABCAAB252352-10X96
- ABCAAB252352-1X384
- Target protein:
- Interferon alpha 1
- Interferon alpha 1
- Interferon alpha 1
- Primary antibody reactivity:
- Mouse
- Mouse
- Mouse
- Assay Type:
- Sandwich
- Sandwich
- Sandwich
- Format:
- Pre-coated
- Pre-coated
- Pre-coated
- Assay duration:
- One step
- One step
- One step
- Sensitivity:
- 7 pg/ml
- 7 pg/ml
- 7 pg/ml
- Detection range:
- 15,6 - 1000 pg/ml
- 15,6 - 1000 pg/ml
- 15,6 - 1000 pg/ml
- Detection method:
- Colorimetric
- Colorimetric
- Colorimetric
- Sample type:
- Cell culture media, Heparin Plasma, Citrate plasma, Serum
- Cell culture media, Heparin Plasma, Citrate plasma, Serum
- Cell culture media, Heparin Plasma, Citrate plasma, Serum
- Time to Results:
- 1 h 30 min
- 1 h 30 min
- 1 h 30 min
- Regulatory status:
- RUO
- RUO
- RUO
- Storage temperature:
- 4 °C
- 4 °C
- 4 °C
Specifications
Product Family Options
Product Information
- Tests per kitAvailabilityPrice
- ABCAAB252352-1X96Mouse Interferon alpha 1 ELISA kitSpecifications:
More
Specifications
Cat. No.ABCAAB252352-1X96Target proteinInterferon alpha 1Primary antibody reactivityMouseAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity7 pg/mlDetection range15,6 - 1000 pg/mlDetection methodColorimetricSample typeCell culture media, Heparin Plasma, Citrate plasma, SerumTime to Results1 h 30 minRegulatory statusRUOStorage temperature4 °C - ABCAAB252352-10X96Mouse Interferon alpha 1 ELISA kitSpecifications:
More
Specifications
Cat. No.ABCAAB252352-10X96Target proteinInterferon alpha 1Primary antibody reactivityMouseAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity7 pg/mlDetection range15,6 - 1000 pg/mlDetection methodColorimetricSample typeCell culture media, Heparin Plasma, Citrate plasma, SerumTime to Results1 h 30 minRegulatory statusRUOStorage temperature4 °C - ABCAAB252352-1X384Mouse Interferon alpha 1 ELISA kitSpecifications:
More
Specifications
Cat. No.ABCAAB252352-1X384Target proteinInterferon alpha 1Primary antibody reactivityMouseAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity7 pg/mlDetection range15,6 - 1000 pg/mlDetection methodColorimetricSample typeCell culture media, Heparin Plasma, Citrate plasma, SerumTime to Results1 h 30 minRegulatory statusRUOStorage temperature4 °C



