Anti-Malaria ELISA Kit
Catalog # ABCAAB178649-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Anti-Malaria ELISA kit
Specifications
About this item
Anti-Malaria ELISA kit is a Indirect ELISA kit for the measurement of Anti-Malaria in Human in Citrate plasma, Heparin plasma, serum samples.