Human PDGF-CC ELISA Kit
Catalog # ABCAAB234568-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human PDGF-CC ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human PDGF-CC ELISA Kit is a single-wash 90-min Simplestep used to quantify Human PDGF-CC with a sensitivity of 2,79 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.