Mouse IL-1 beta Matched Antibody Pair Kit
ABCAAB210895-5X96
: Abcam
:
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:IL-1 beta
- Description:Mouse IL-1 beta Matched antibody pair kit
- Detection method:Colorimetric
- Detection range:7,8 - 500 pg/ml
- Storage temperature:4 °C
- Sensitivity:1,7 pg/ml
- Regulatory status:RUO
- Tests per kit:5×96 tests
Mouse IL-1 beta Matched antibody pair kit is a kit for ELISA development for the measurement of Mouse IL-1 beta by sandwich ELISA method.