Cotinine ELISA Kit
Catalog # ABCAAB285286-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Cotinine ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Cotinine ELISA Kit is a Competitive ELISA kit for the measurement of Cotinine in Human in Urine, Serum samples.