Anti-Ramucirumab ELISA Kit
ABCAAB282899-1X96
: Abcam
:
- Description:Anti-Ramucirumab ELISA Kit
- Tests per kit:96 tests
Anti-Ramucirumab ELISA Kit is an anti-drug antibody (ADA) ELISA used for the qualitative determination of antibodies to Ramucirumab - Colorimetric ELISA - 450 nm readout - works on any plate reader - Positive and Negative controls included.