Human IL-12 p70 ELISA Kit
Supplier: Abcam
Certificates
About this item
Human IL-12 p70 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-12 p70 in Human in plasma, cell culture supernatant, serum samples.
Human IL-12 p70 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-12 p70 in Human in plasma, cell culture supernatant, serum samples.