Human IL-6 ELISPOT Kit
Catalog # ABCAAB62936-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-6 ELISPOT kit
- Species reactivity:Human
- Form:Pre-coated
- Storage temperature:4 °C
- Processing time:Multiple steps
- Antigen symbol:IL-6
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human IL-6 ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Human IL-6 production in a single cell suspension.