Order Entry
Export
Orders LinkContactUsLinkComponent
Human VCAM1 ELISA Kit

Human VCAM1 ELISA Kit

Human VCAM1 ELISA kit, 96 tests
Catalog # ABCAAB223591-96
Supplier:   Abcam
Human VCAM1 ELISA Kit
Catalog # ABCAAB223591-96
Supplier:  Abcam
CAS Number:  
Human VCAM1 ELISA Kit
Human VCAM1 ELISA Kit

Human VCAM1 ELISA kit, 96 tests

CAS Number:  
Price
749,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human VCAM1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human VCAM1 with a sensitivity of 37 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader. - Cited in over 10 citations.

Specifications

Specifications

  • Description:
    Human VCAM1 ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
  • Detection range:
    0,14 - 9 ng/ml
  • Storage temperature:
    +4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB223591-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    37 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    VCAM1
  • Regulatory status:
    RUO

Specifications