Mouse GDF-15 ELISA Kit
Catalog # ABCAAB216947-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse GDF-15 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse GDF-15 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse GDF-15 with a sensitivity of 22,2 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.