Mouse Neurotrophin-3 ELISA Kit
Catalog # ABCAAB213882-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Neurotrophin-3 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Neurotrophin-3 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Neurotrophin-3 in Mouse in Cell culture supernatant, Serum samples.