Nivolumab ELISA Kit
Catalog # ABCAAB237651-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Nivolumab ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Nivolumab ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Nivolumab in Human in Plasma, Serum samples.