HSP70 ELISA Kit
Catalog # ABCAAB133060-96
Supplier: Abcam
CAS Number:
Specifications
- Description:HSP70 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
HSP70 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of HSP70 in Mouse, Rat, Human in Tissue Extracts, Cell Lysate samples.