Vedolizumab ELISA Kit
Catalog # ABCAAB264503-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Vedolizumab ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Vedolizumab ELISA Kit is a Quantitative ELISA kit for the measurement of Vedolizumab in Human in Plasma, Serum samples.