Order Entry
Export
ContactUsLinkComponent
Rat Kallikrein 10 ELISA Kit
Rat Kallikrein 10 ELISA Kit
Catalog # ABCAAB284619-1X96
Supplier:  Abcam
CAS Number:  
undefined
Rat Kallikrein 10 ELISA Kit
Catalog # ABCAAB284619-1X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Rat Kallikrein 10 ELISA kit
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Rat Kallikrein 10 ELISA kit is a single-wash 90-min Simplestep used to quantify Rat Kallikrein 10 with a sensitivity of 2,35 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.