Order Entry
Export
ContactUsLinkComponent
 
Human IL-17F ELISpot Kit
 :  Abcam
 
undefined
Human IL-17F ELISpot Kit
 :  Abcam

 

  • Species reactivity:
    Human
  • Sample type:
    Suspension cells
  • Form:
    Pre-coated
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Antigen symbol:
    IL-17F
  • Regulatory status:
    RUO