Order Entry
Export
Orders LinkContactUsLinkComponent
Human Factor B ELISA Kit

Human Factor B ELISA Kit

Human Factor B ELISA kit, 96 tests
Catalog # ABCAAB300318-96
Supplier:   Abcam
Human Factor B ELISA Kit
Catalog # ABCAAB300318-96
Supplier:  Abcam
CAS Number:  
Human Factor B ELISA Kit
Human Factor B ELISA Kit

Human Factor B ELISA kit, 96 tests

CAS Number:  
Price
795,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human Factor B ELISA kit is a single-wash 90-min SimpleStep ELISA® for the quantitative measurement of Human Factor B in cell culture supernatant, EDTA plasma, heparin plasma, milk, saliva serum, urine samples.

Specifications

Specifications

  • Description:
    Human Factor B ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Cell culture supernatant, EDTA Plasma, Heparin Plasma, Milk, Saliva, Serum, Urine
  • Detection range:
    53,125 - 3400 pg/ml
  • Storage temperature:
    4 ℃
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB300318-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    18,18 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Factor B
  • Regulatory status:
    RUO

Specifications