Order Entry
Export
Orders LinkContactUsLinkComponent
Human CD63 ELISA Kit

Human CD63 ELISA Kit

Supplier:   Abcam
Human CD63 ELISA Kit
Supplier:  Abcam
Certificates
Human CD63 ELISA Kit

2 Options Available Below

Product Details & Documents

Human CD63 ELISA kit is a single-wash 90-min Simplestep used to quantify Human CD63 with a sensitivity of 33 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCAAB275099-1X96
  • ABCAAB275099-1X384
  • Target protein:
  • CD63
  • CD63
  • Primary antibody reactivity:
  • Human
  • Human
  • Assay Type:
  • Sandwich
  • Sandwich
  • Format:
  • Pre-coated
  • Pre-coated
  • Assay duration:
  • One step
  • One step
  • Sensitivity:
  • 33 pg/ml
  • 33 pg/ml
  • Detection range:
  • 46,9 - 3000 pg/ml
  • 46,9 - 3000 pg/ml
  • Detection method:
  • Colorimetric
  • Colorimetric
  • Sample type:
  • Cell culture supernatant, Serum, EDTA Plasma
  • Cell culture supernatant, Serum, EDTA Plasma
  • Time to Results:
  • 1 hr 30 min
  • 1 hr 30 min
  • Regulatory status:
  • RUO
  • RUO
  • Storage temperature:
  • 4 °C
  • 4 °C

Specifications

Product Family Options

Product Information

  • Tests per kit
    Availability
    Price
  • ABCAAB275099-1X96
    Human CD63 ELISA Kit
    Human CD63 ELISA Kit
    96 tests
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 723,00 €
    723,00 €
    1 * 1 KIT
       723,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB275099-1X96
    Target protein
    CD63
    Primary antibody reactivity
    Human
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    One step
    Sensitivity
    33 pg/ml
    Detection range
    46,9 - 3000 pg/ml
    Detection method
    Colorimetric
    Sample type
    Cell culture supernatant, Serum, EDTA Plasma
    Time to Results
    1 hr 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
  • ABCAAB275099-1X384
    Human CD63 ELISA Kit
    Human CD63 ELISA Kit
    384 tests
    Log in for availability
    Log in for availability
    1 * 384 Tests
    1 * 384 Tests - 1 860,00 €
    1 860,00 €
    1 * 384 Tests
       1 860,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB275099-1X384
    Target protein
    CD63
    Primary antibody reactivity
    Human
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    One step
    Sensitivity
    33 pg/ml
    Detection range
    46,9 - 3000 pg/ml
    Detection method
    Colorimetric
    Sample type
    Cell culture supernatant, Serum, EDTA Plasma
    Time to Results
    1 hr 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C