Pig IL-8 ELISA Kit
Catalog # ABCAAB113352-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Pig IL-8 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Pig IL-8 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Pig IL-8 in Pig in plasma, cell culture supernatant, serum samples.