Androstenedione ELISA Kit
Catalog # ABCAAB108672-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Androstenedione ELISA kit
Specifications
About this item
Androstenedione ELISA kit is a Competitive ELISA kit for the measurement of Androstenedione in Citrate plasma, Heparin plasma, serum samples.