Human IL-36 alpha ELISA (IL-1F6)
Catalog # ABCAAB267641-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-36 alpha ELISA (IL-1F6)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Human IL-36 alpha ELISA (IL-1F6) is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-36 alpha ELISA (IL-1F6) in Human in cell culture supernatant, citrate plasma, EDTA plasma, serum samples.