Human 14-3-3 tau ELISA Kit
Catalog # ABCAAB246543-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human 14-3-3 tau ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human 14-3-3 tau ELISA kit is a single-wash 90-min Simplestep used to quantify Human 14-3-3 tau with a sensitivity of 6,1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.