Mouse MCP5 ELISA Kit
ABCAAB100723-96
: Abcam
:
- Description:Mouse MCP5 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Mouse MCP5 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse MCP5 in Mouse in plasma, cell culture supernatant, serum samples.