Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse PSAP ELISA Kit

Mouse PSAP ELISA Kit

Mouse PSAP ELISA Kit, 96 tests
Catalog # ABCAAB315297-96TST
New Product
Supplier:   Abcam
Mouse PSAP ELISA Kit
Catalog # ABCAAB315297-96TST
Supplier:  Abcam
New Product
CAS Number:  
Mouse PSAP ELISA Kit
Mouse PSAP ELISA Kit

Mouse PSAP ELISA Kit, 96 tests

CAS Number:  
Price
692,00 €
Log in for availability
1 * 1 Tests
Product Details & Documents

Mouse PSAP ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse PSAP with a sensitivity of 13.4 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifications

Specifications

  • Description:
    Mouse PSAP ELISA Kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, EDTA Plasma, Citrate plasma, Heparin Plasma, Cell culture supernatant
  • Detection range:
    43,75 - 2800 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 hr 30 min
  • Cat. No.:
    ABCAAB315297-96TST
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    13,4 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    PSAP
  • Regulatory status:
    RUO

Specifications