ContactUsLinkComponent
Human GDF-15 ELISA Kit
Human GDF-15 ELISA Kit
Catalog # ABCAAB155432-96
Supplier:  Abcam
CAS Number:  
undefined
Human GDF-15 ELISA Kit
Catalog # ABCAAB155432-96
Supplier:  Abcam
CAS Number:  
About this item
Human GDF-15 ELISA kit ab155432 is used to quantify Human GDF-15 with a sensitivity of 2 pg/ml. Sandwich ELISA kit.