Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse Transferrin ELISA Kit

Mouse Transferrin ELISA Kit

Mouse Transferrin ELISA kit, 96 tests
Catalog # ABCAAB157724-96
Supplier:   Abcam
Mouse Transferrin ELISA Kit
Catalog # ABCAAB157724-96
Supplier:  Abcam
CAS Number:  
Mouse Transferrin ELISA Kit
Mouse Transferrin ELISA Kit

Mouse Transferrin ELISA kit, 96 tests

CAS Number:  
Price
790,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse Transferrin ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Transferrin in Mouse in plasma, Serum samples.

Specifications

Specifications

  • Description:
    Mouse Transferrin ELISA kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB157724-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,6935 ng/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Plasma, serum
  • Target protein:
    Transferrin
  • Detection range:
    3,13 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Assay Type:
    Sandwich

Specifications